D(+)-Methioninol

Lieferant: Molekula

Synonyme: D-Methioninol

M86575548
MOLEM86575548EA 0 EUR
MOLEM86575548
D(+)-Methioninol
D(+)-Methioninol
Formel: C₅H₁₃NOS
Molecular Weight: 135,2 g/mol
CAS-Nummer: 87206-44-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR