(Ethoxycarbonylmethyl)triphenylphosphoniumbromid
Lieferant: Molekula
Synonyme:
(Carbethoxymethyl)triphenylphosphonium bromide, (2-Ethoxy-2-oxoethyl)triphenylphosphonium bromide
M87142634
MOLEM87142634EA
0
EUR
MOLEM87142634
(Ethoxycarbonylmethyl)triphenylphosphoniumbromid
(Ethoxycarbonylmethyl)triphenylphosphoniumbromid
Formel:
C₂₂H₂₂BrO₂P Molecular Weight: 429,30 g/mol |
MDL:
MFCD00011835 CAS-Nummer: 1530-45-6 EINECS: 216-230-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...