Dapson
Lieferant: Molekula
|
Achtung
|
Synonyme:
DDS, 4-Aminophenylsulfon, Bis(4-aminophenyl)sulfon, 4,4'-Sulfonyldianilin, 4,4'-Diaminodiphenylsulfon
M87192589
MOLEM87192589EA
0
EUR
MOLEM87192589
Dapson
Dapson
Formel:
C₁₂H₁₂N₂O₂S Molecular Weight: 248,31 g/mol |
MDL:
MFCD00007887 CAS-Nummer: 80-08-0 EINECS: 201-248-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...