6-Amino-m-kresol
Lieferant: Molekula
Synonyme:
2-Amino-5-methylphenol, 2-Hydroxy-4-methylanilin, 1-Amino-2-hydroxy-4-methylbenzol, 4-Amino-3-hydroxytoluol
M87204758
MOLEM87204758EA
0
EUR
MOLEM87204758
6-Amino-m-kresol
6-Amino-m-kresol
Formel:
C₇H₉NO Molecular Weight: 123,15 g/mol Siedepunkt: 259 °C (1013 hPa) Schmelzpunkt: 162 °C |
MDL:
MFCD00007693 CAS-Nummer: 2835-98-5 EINECS: 220-620-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...