5-Chloruracil
Lieferant: Molekula
Synonyme:
5-Chlor-2,4-dihydroxypyrimidin, Chloruracil
M87447286
MOLEM87447286EA
0
EUR
MOLEM87447286
5-Chloruracil
5-Chloruracil
Formel:
C₄H₃ClN₂O₂ Molecular Weight: 146,53 g/mol Schmelzpunkt: > 300 °C |
MDL:
MFCD00006019 CAS-Nummer: 1820-81-1 EINECS: 217-339-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...