4-Chinolincarbaldehyd
Lieferant: Molekula
Synonyme:
4-Chinolincarboxaldehyd
M87482000
MOLEM87482000EA
0
EUR
MOLEM87482000
4-Chinolincarbaldehyd
4-Chinolincarbaldehyd
Formel:
C₁₀H₇NO Molecular Weight: 157,17 g/mol Siedepunkt: 122…123 °C (4 mmHg) Schmelzpunkt: 150 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00006781 CAS-Nummer: 4363-93-3 EINECS: 224-453-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...