Amantadin Hydrochlorid

Lieferant: Molekula

Synonyme: 1-Adamantylamin Hydrochlorid, 1-Aminotricyclo[3.3.1.1(3,7)]decan Hydrochlorid, Tricyclo[3.3.1.1(3,7)]dec-1-ylaminhydrochlorid, 1-Adamantanammoniumchlorid, 1-Aminoadamantan Hydrochlorid

M87836477
MOLEM87836477EA 0 EUR
MOLEM87836477
Amantadin Hydrochlorid
Amantadin Hydrochlorid
Formel: C₁₀H₁₈ClN
Molecular Weight: 187,71 g/mol
Schmelzpunkt: 360 °C
MDL: MFCD00074723
CAS-Nummer: 665-66-7
EINECS: 211-560-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR