2-Chlornicotinonitril
Lieferant: Molekula
Synonyme:
2-Chlor-3-pyridincarbonsäurenitril, 2-Chlornicotinsäurenitril, 2-Chlor-3-pyridincarbonitril, 2-Chlor-3-cyanpyridin
M87915489
MOLEM87915489EA
0
EUR
MOLEM87915489
2-Chlornicotinonitril
2-Chlornicotinonitril
Formel:
C₆H₃ClN₂ Molecular Weight: 138,56 g/mol Siedepunkt: 112 °C (1 mmHg) Schmelzpunkt: 107…108 °C |
MDL:
MFCD00014628 CAS-Nummer: 6602-54-6 EINECS: 229-550-1 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...