α-Jonon

Lieferant: Molekula

Synonyme: (E)-4-(2,6,6-Trimethylcyclohex-2-en-1-yl)but-3-en-2-one

M87966374
MOLEM87966374EA 0 EUR
MOLEM87966374
α-Jonon
α-Jonon
Formel: C₁₃H₂₀O
Molecular Weight: 192,3 g/mol
MDL: MFCD00001565
CAS-Nummer: 127-41-3
EINECS: 204-841-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR