Chlorphenolrot
Lieferant: Molekula
Synonyme:
4,4'-(3H-2,1-Benzoxathiol-3-yliden)bis[2-chlorphenol]-S,S-dioxid, 3′,3′′-Dichlorphenolsulfonphthalein
M88339115
MOLEM88339115EA
0
EUR
MOLEM88339115
Chlorphenolrot
Chlorphenolrot
Formel:
C₁₉H₁₂Cl₂O₅S Molecular Weight: 423,27 g/mol Schmelzpunkt: ∼261 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005877 CAS-Nummer: 4430-20-0 EINECS: 224-619-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...