3-Amino-2-pyrazincarbonsäure

Lieferant: Molekula

Synonyme: 3-Amino-2-pyrazinecarboxylic acid

M88382615
MOLEM88382615EA 0 EUR
MOLEM88382615
3-Amino-2-pyrazincarbonsäure
3-Aminopyrazine-2-carboxylic acid
Formel: C₅H₅N₃O₂
MDL: MFCD00006141
CAS-Nummer: 5424-01-1
EINECS: 226-558-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR