(2E,4E)-2,4-Hexadienal

Lieferant: Molekula

M88403233
MOLEM88403233EA 0 EUR
MOLEM88403233
(2E,4E)-2,4-Hexadienal
trans,trans-2,4-Hexadienal
Formel: C₆H₈O
Molecular Weight: 96,13 g/mol
MDL: MFCD00007004
CAS-Nummer: 142-83-6
EINECS: 205-564-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR