2-(4-Aminophenyl)ethanol
Lieferant: Molekula
Synonyme:
4-Aminophenethylalkohol, 2-(4-Aminophenyl)ethylalkohol
M89211937
MOLEM89211937EA
0
EUR
MOLEM89211937
2-(4-Aminophenyl)ethanol
2-(4-Aminophenyl)ethanol
Formel:
C₈H₁₁NO Molecular Weight: 137,18 g/mol Siedepunkt: 235…255 °C (1013 hPa) Schmelzpunkt: 108 °C |
MDL:
MFCD00007922 CAS-Nummer: 104-10-9 EINECS: 203-174-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...