7-Bromindol

Lieferant: Molekula
Achtung

Synonyme: 7-Bromo-1H-indole

M89512160
MOLEM89512160EA 0 EUR
MOLEM89512160
7-Bromindol
7-Bromindol
Formel: C₈H₆BrN
Molecular Weight: 196,05 g/mol
MDL: MFCD00799492
CAS-Nummer: 51417-51-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR