4-Chlorpyridin Hydrochlorid

Lieferant: Molekula

Synonyme: 4-Chloropyridinium chloride

M89759763
MOLEM89759763EA 0 EUR
MOLEM89759763
4-Chlorpyridin Hydrochlorid
4-Chlorpyridin Hydrochlorid
Formel: C₅H₅Cl₂N
MDL: MFCD00012829
CAS-Nummer: 7379-35-3
EINECS: 230-946-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR