Thiophen-2-acetonitril
Lieferant: Molekula
Synonyme:
Thiophene-2-acetonitrile, 2-Thiopheneacetonitrile, 2-(Thiophen-2-yl)acetonitrile
M89821532
MOLEM89821532EA
0
EUR
MOLEM89821532
Thiophen-2-acetonitril
Thiophen-2-acetonitril
Formel:
C₆H₅NS Molecular Weight: 123,18 g/mol |
MDL:
MFCD00005453 CAS-Nummer: 20893-30-5 EINECS: 244-104-6 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...