The Moticam 1080X, designed for ease of use and the ability to output fast live images in Full HD (1920x1080px), displays your live images on a screen by connecting the camera through the HDMI port.
Make live measurements, notes, image adjustments and more, thanks to its on-board software that can be controlled through the screen by connecting a USB mouse to the camera. By having an integrated SD card slot, saving images will not require a computer, making it easy to save and organise your images and videos. Its Wi-Fi connectivity allows you to connect with a computer or mobile device, even simultaneously, and share your live images with ease.
Software: With its own built-in software, the Moticam 1080X makes it possible to view live images, make measurements, image adjustments, or edit them. Images or videos are captured directly from a screen with mouse, storing them in an SD card.
Functions: Including exposure, gain, white balance, color adjustment, sharpness and denoising control, zoom, mirror, comparison, freeze, measure, cross, browse function.
Lieferumfang: CS ring adaptor, focusable lens, 30 and 38mm eyepiece adapters, Motic 4-dot calibration slide, macro calibration card, macro tube, universal power supply, HDMI cable, Wi-Fi dongle and USB Pendrive with Motic Images Plus 3.1.