Moticam 1080X HDMI Camera
Lieferant: MOTIC
1100600101481
MOTI1100600101481EA 1253 EUR
MOTI1100600101481
Moticam 1080X HDMI Camera
Kameras
The Moticam 1080X, designed for ease of use and the ability to output fast live images in Full HD (1920x1080px), displays your live images on a screen by connecting the camera through the HDMI port.
Make live measurements, notes, image adjustments and more, thanks to its on-board software that can be controlled through the screen by connecting a USB mouse to the camera. By having an integrated SD card slot, saving images will not require a computer, making it easy to save and organise your images and videos. Its Wi-Fi connectivity allows you to connect with a computer or mobile device, even simultaneously, and share your live images with ease.
Software: With its own built-in software, the Moticam 1080X makes it possible to view live images, make measurements, image adjustments, or edit them. Images or videos are captured directly from a screen with mouse, storing them in an SD card.
Functions: Including exposure, gain, white balance, color adjustment, sharpness and denoising control, zoom, mirror, comparison, freeze, measure, cross, browse function.
Lieferumfang: CS ring adaptor, focusable lens, 30 and 38mm eyepiece adapters, Motic 4-dot calibration slide, macro calibration card, macro tube, universal power supply, HDMI cable, Wi-Fi dongle and USB Pendrive with Motic Images Plus 3.1.