Full face mask, Advantage 3100
Lieferant: M S A
10027725 10027724 10027723
MSAA10027725EA 243 EUR
MSAA10027725 MSAA10027724 MSAA10027723
Full face mask, Advantage 3100
Atemschutzmasken Mehrweg-Atemschutzmasken Vollmasken
The Advantage 3100 full mask series provides high protection and unique comfort. The soft sealing line made from hypoallergenic silicone provides a pressure free fit.
  • Large, optically corrected lens ensures a clear, undistorted view
  • Grey-blue colour gives the mask an aesthetic appearance
  • Quick, simple donning without painful hair pulling
  • Patented head harness adapts perfectly to the facial contours
Suitable for use with all filter series with wire connection according to EN 148.
Standards: EN 136 class 3