Extraction systems and accessories for acid storage cabinets
Lieferant: asecos
10147 10148 10149 10150 10151 10152 10153 10155 10157 10158 10162 10163 10164 10177 10178 10195 10196 10197 10154
MSGA10147EA 355 EUR
MSGA10147 MSGA10148 MSGA10149 MSGA10150 MSGA10151 MSGA10152 MSGA10153 MSGA10155 MSGA10157 MSGA10158 MSGA10162 MSGA10163 MSGA10164 MSGA10177 MSGA10178 MSGA10195 MSGA10196 MSGA10197 MSGA10154
Extraction systems and accessories for acid storage cabinets
Möbel Lagerschränke
Powder coated steel tube with wall mount and flat hoods. Can be easily fastened onto workbenches, tables. It can be easily adjustable has suction valve with swivel function.
  • Extremely light
  • High position stability and moving sensitivity