Trolley for gas cylinders
Lieferant: asecos
13267
MSGA13267EA 428 EUR
MSGA13267
Trolley for gas cylinders
Transportwagen
This trolley made of PE can equip up to 4 cylinders with a load capacity of 300 kg. Safe transport is ensured with the provision for forklift.
Lieferumfang: With lashing straps and safety chain.