Tipping skips
Lieferant: asecos
23080 23082 23083 23084 23086 23087 23088 23090 23091 23092 23094 23095 13061 13062 13063 13064 15489 15490 15492 15493 15495 15496 15498 15499
MSGA23080EA 777 EUR
MSGA23080 MSGA23082 MSGA23083 MSGA23084 MSGA23086 MSGA23087 MSGA23088 MSGA23090 MSGA23091 MSGA23092 MSGA23094 MSGA23095 MSGA13061 MSGA13062 MSGA13063 MSGA13064 MSGA15489 MSGA15490 MSGA15492 MSGA15493 MSGA15495 MSGA15496 MSGA15498 MSGA15499
Tipping skips
Transportwagen
Tipping skips are equipped with subframe with inserts for transportation by forklift truck. Releasing the locking device immediately activates the tipping process, with integrated spring mechanism limits the tipping process to ∼90° and prevents the container of being damaged.
  • Easy to move due to large castors (Ø 200mm) and swivel castors (Ø 125mm) of polyamide, lockable
  • Easy emptying by tipping the container over the axle of the wheels
  • Extremely sturdy, seamless skips
  • Easy handling and easy to clean
  • Impervious to fluids and resistant against acids and alkalis
  • Integrated underframe with inserts
Made from polyethylene - provision of small parts or separate collection of waste and recyclable materials.