Ultra fine absorption grains
Lieferant: asecos
14394 18165
MSGA14394EA 31 EUR
MSGA14394 MSGA18165
Ultra fine absorption grains
Absorptionsmaterial
Fine absorption granules for oil, alkalis, grease, motor fuels, varnishes.
  • Officially tested and approved for use on traffic areas
  • 0,3 to 0,7 mm granularity
  • 10 kg sack binds up to 11 litres of heating oil
  • Fills smallest cracks and openings
  • Developed for special purposes where a minimum of waste and a maximum of absorption is needed