Safety storage cabinets, XL-Classic-90
Lieferant: asecos
30382001
MSGA30382001EA 5784 EUR
MSGA30382001
Safety storage cabinets, XL-Classic-90
Möbel Lagerschränke
Fireproof safety cabinets, classified type 90 (fire resistant up to 90 minutes).
  • Slide bar door closer
  • Safety elements assembled outside the storage compartment for increased protection against corrosion
  • Door open arrest system (optional), convenient and safe removal or storage of containers
  • Doors lockable with cylinder locking (integration in a locking system possible)
  • Integrated air ducts ready for connection (NW 75) to a technical exhaust system
EN 14727 conformity (laboratory furniture), GS approval, CE conformity. Furnace tested (type test) in accordance with EN 14470-1
Standards: EN 14727 conformity (laboratory furniture), GS approval, CE conformity. Furnace tested (type test) in accordance with EN 14470-1
Lieferumfang: Supplied without interior equipment packages. Interior equipment packages need to be ordered in addition.