Cabinet for chemicals for safe storage of non - flammable chemicals and toxic substances
Lieferant: asecos
30493041 30495041
MSGA30493041EA 1065 EUR
MSGA30493041 MSGA30495041
Cabinet for chemicals for safe storage of non - flammable chemicals and toxic substances
Möbel Lagerschränke
Safe and proper storage of non-flammable chemicals and toxic substances in working areas.
  • Robust construction and longevity: robust, high-quality, scratch-resistant outer body with powder-coated textured surface
  • No unauthorised use: doors lockable with cylinder locking (integrated in the turning handle)
  • Easy alignment: adjusting aids to compensate for uneven floor
Model CS-CLASSIC, CS.195.105. with wing doors.
Model CS-CLASSIC-G, CS.195.105.WDFW with glass-panelled wing doors.
Ventilation: natural ventilation due to ventilation openings at theMore Product Information
Bestellen
TECHNISCHE DATEN
Environmentally Preferable