Environmental safety cabinets, E-Classic
Lieferant: asecos
30628007
MSGA30628007EA 1106 EUR
MSGA30628007
Environmental safety cabinets, E-Classic
Möbel Lagerschränke
Environmental cabinets are ideal for a safe and approved storage of water pollutant liquids in working areas.
  • Sheet-steel construction
  • Tall cabinet with wing doors
  • Robust construction and longevity
  • With cylinder locking (integrated in turning handle)
Bestellinformation: Interior equipment packages need to be ordered separately (not included). Please check the website or contact Avantor for further information.