Hazardous material work station, EX-LINE GAPEX model with robust construction made of chemically resistant, anodised aluminium profiles.
- Required occupational exposure limits are maintained
- used for testing of the air equipment according to EN 14175 Part 3 (5.4.4)
- Maximum user safety
- Transparent side panes made from 5 mm (toughened safety glass)
- Highly effective capturing of hazardous materials inside the work station through special fresh air curtain technology
- Prepared for connection to the on-site exhaust air system
Standard equipment consists of:
Control box with complete monitoring electronics incl. potential-free switching contact, e.g. for controlling on-site exhaust air system (must be installed outside the Ex zone) - Cable length 5 m
Transparent side panes made from 5 mm (toughened safety glass)
Fold-away deflector
Melamine resin coated rear wall
Working surface made from stainless steel (V2A/1.4301) as seal welded sump
Support frame for standing work
Earthing link, mounted in the profile
EX-LED lamp 6500 K
Standards: Electrical equipment approved in accordance with 2014/34/EU.