Safety Storage Cabinet, Type 90, ION-PRO-90 CHARGE model IO90.195.120.PC.WDC
Lieferant: asecos
41269047 41391 41392 41393 41394
135-2884EA 6703 EUR
135-2884 MSGA41391 MSGA41392 MSGA41393 MSGA41394
Safety Storage Cabinet, Type 90, ION-PRO-90 CHARGE model IO90.195.120.PC.WDC
Möbel Lagerschränke
Safe active and passive storage of lithium-ion batteries.
  • Robust construction and longevity; Convenient cabinet entry
  • Integrated potential-free alarm contact; No unauthorised use
  • Integrated transport base; Easy alignment
  • Smoke detector and temperature sensor; Extraction unit
  • Pressure relief; All-around protection
  • LED display
  • Fire suppression system
Robust construction and longevity: robust, high-quality, scratch-resistant outer body with powder-coated textured surface. Convenient cabinet entry: open both doors at the same time with just one hand.More Product Information