Oil sorbents mats
Lieferant: asecos
11327 9934
MSGA11327EA 139.5 EUR
MSGA11327 MSGA9934
Oil sorbents mats
Absorptionsmaterial
Water repellent mats for abosrption and clear up of oils and petrol based liquid products.
  • Capacity up to 16 times of its own weight
  • High rib and rub resistance
  • Perforated to only take sufficient material as required