Universal socks
Lieferant: asecos
11347 9954
MSGA11347EA 121.5 EUR
MSGA11347 MSGA9954
Universal socks
Absorptionsmaterial
Universal socks for safe absorption of oil, water, cooling liquids and non aggressive chemicals.
  • Optimal for long term use
  • Capacity up to 16 times of its own weight
  • High rib and rub resistance
For containment and absorption of larger leaks.