D(-)-Luciferin

Lieferant: MARKET SOURCE PART PROCESS

Synonyme: D-Luciferin, (S)-2-(6-Hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid

102111
MSPP102111EA 0 EUR
MSPP102111
D(-)-Luciferin
D(-)-Luciferin
Formel: C₁₁H₈N₂O₃S₂
Molecular Weight: 280,3228 g/mol
MDL: MFCD00042929
CAS-Nummer: 2591-17-5
EINECS: 219-981-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR