Brij® 35 30% in wässriger Lösung

Lieferant: MARKET SOURCE PART PROCESS

Synonyme: Brij® L23

13900
MSPP13900EA 98.5 EUR
MSPP13900
Brij® 35 30% in wässriger Lösung
Brij® 35
Formel: C₅₈H₁₁₈O₂₄
Molecular Weight: 1199.56 g/mol
MDL: MFCD00080891
CAS-Nummer: 9002-92-0
EINECS: 500-002-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR