Adenosin-5'-diphosphorsäure (ADP)
Lieferant: MARKET SOURCE PART PROCESS
Synonyme:
Adenosine 5'-diphosphate, 5′-ADP, Adenosine 5′-diphosphate
CH384
MSPPCH384EA
0
EUR
MSPPCH384
Adenosin-5'-diphosphorsäure (ADP)
Adenosin-5'-diphosphorsäure (ADP)
Formel:
C₁₀H₁₅N₅O₁₀P₂ Molecular Weight: 427.2 g/mol |
MDL:
MFCD00066473 CAS-Nummer: 58-64-0 EINECS: 200-392-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...