Silicone rubber sponge stoppers
Lieferant: CHEMGLASS
CLS-1490-028 CLS-1490-038
CGLACLS-1490-028EA 0 EUR
CGLACLS-1490-028 MSPPCLS-1490-038
Silicone rubber sponge stoppers
Stopfen Silikonstopfen
This silicone sponge closure is manufactured with a sealed in silicone sponge filter. Flange fits snugly over the necks of vessels permitting the exchange of gases, while reducing evaporation and preventing the passing of aerosols.
  • Ideal for use with aerobic cultures
  • Reusable and autoclavable
  • Withstand heat sterilisation up to 180 °C