6-Brom-1-hexanol
Lieferant: MARKET SOURCE PART PROCESS
|
Achtung
|
Synonyme:
Hexamethylenbromhydrin, 1-Brom-6-hydroxyhexan
ST01360-1KG
MSPPST01360-1KGEA
0
EUR
MSPPST01360-1KG
6-Brom-1-hexanol
6-Brom-1-hexanol
Formel:
BrH₂C(CH₂)₄CH₂OH Molecular Weight: 181,07 g/mol Siedepunkt: 105 °C (1013 hPa) Dichte: 1,3838 g/cm³ (24 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00002983 CAS-Nummer: 4286-55-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...