Trolleys for Containers
Lieferant: MULTIROIR
65010
MULC65010EA 187 EUR
MULC65010
Trolleys for Containers
Transportwagen
This trolley is unique on the market for its robustness and quality. It is suitable for moving heavy loads and sensitive products.
  • Tray size: 670×460 mm
  • Rigid PP base reinforced with 2 metal bars
  • Non-slip tray
  • Ergonomic grip thanks to the handle
  • Non-staining castors with ball bearings
  • Practical handling hook
  • Trolleys delivered unassembled, easy to assemble the castors
  • Wheel diameter: 75 mm
The range is available in two versions:
Flatbed trolley, suitable for heavy and bulky loads
Bag trolley with 3 safety edges, suitable for all 600×400 Euro containers, for safe transport