Cetrimoniumbromid ≥98,0%, BAKER ANALYZED®, J.T.Baker®
Lieferant: Avantor
|
Achtung
|
Synonyme:
Hexadecyltrimethylammoniumbromid, N-Cetyl-N,N,N-trimethylammoniumbromid, CTAB, N-Hexadecyl-N,N,N-trimethylammoniumbromid, Cetyltrimethylammoniumbromid
N121-07EA
0
EUR
N121-07
Cetrimoniumbromid ≥98,0%, BAKER ANALYZED®, J.T.Baker®
Cetrimoniumbromid
Formel:
CH₃(CH₂)₁₅N(Br)(CH₃)₃ Molecular Weight: 364,45 g/mol Schmelzpunkt: 237…243 °C Flammpunkt: 244 °C (geschlossener Tiegel) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011772 CAS-Nummer: 57-09-0 EINECS: 200-311-3 UN: 3077 ADR: 9,III Merck Index: 13,02034 |
Spezifikation Testergebnisse
For Laboratory,Research,or Manufacturing Use | >= 98.0 % |
Assay (C₁₉H₄₂BrN) | ≥98.0 % |
Insoluble Matter | ≤0.005 % |
Residue after Ignition | ≤0.10 % |
Appearance | Passes Test |
Identification (by IR) | Passes Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...