Neutralrot-Lösung, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: Basisrot 5, 2-Amino-8-(dimethylamino)-3-methylphenaziniumchlorid, N8,N8,3-Trimethyl-2,8-phenazindiamin Monohydrochlorid

N6264-50ML
N6264-50MLEA 141 EUR
N6264-50ML
Neutralrot-Lösung, Sigma-Aldrich®
Neutralrot

Used as a red nuclear counterstain in various cytochemical procedures.

Formel: C₁₅H₁₇ClN₄
Molecular Weight: 288,78 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00012651
CAS-Nummer: 553-24-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a45ed674ccf6293f0268c049f029572c9a5ecbaa&vid=a45ed674ccf6293f0268c049f029572c9a5ecbaa&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR