Kernechtrot, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: Natrium-1-amino-2,4-dihydroxy-9,10-dihydro-9,10-dioxoanthracen-3-sulfonat, Heliechtrubin

M-Clarity™ Qualitätsstufe = MQ200

N8002-5G
N8002-5GEA 194 EUR
N8002-5G
Kernechtrot, Sigma-Aldrich®
Kernechtrot

Nuclear Fast Red is suitable for the determination of calcium. It is suitable for use in prussian blue staining in neural progenitor cells. It may be employed as highly sensitive 'off/on' fluorescent probe for the determination of guanine. Nuclear fast red (2-anthracenesulfonic acid, 4-amino-9,10-dihydro-1,3-dihydroxy-9,10-dioxo-, monosodium salt) may be used as selective complexing agent for the simultaneous determination of copper and bismuth by adsorptive stripping voltammetry.


  • Used as a biological stain

Formel: C₁₄H₈NNaO₇S
Molecular Weight: 357,28 g/mol
Schmelzpunkt: 300 °C
Lagertemperatur: Room Temperature
MDL: MFCD00078493
CAS-Nummer: 6409-77-4
EINECS: 229-088-0
Merck Index: 13,06512
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5f353df3de5fa370a6fcdd2551f9bf7e2434a30&vid=c1c6ebeb11dc30427146c0ba6daaebbe718df049&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Dark Brown to Very Dark Brown and Dark
Appearance (Form) Powder
Solubility (Colour)
c = 4 mg/4ml; Water
Dark Red to Very Dark Red
Suitability
Suitable for Determination of Calcium
Suitable
Recommended Retest Period 4 years

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR