XTT Natrium Salz, hochrein

Lieferant: VWR Chemicals

Synonyme: 2,3-Bis(2-methoxy-4-nitro-5-sulphophenyl)-2H-tetrazolium-5-carboxanilide, XTT sodium, Sodium 5,5'-(5-(phenylcarbamoyl)-2H-tetrazole-3-ium-2,3-diyl)bis(4-methoxy-2-nitrobenzenesulphonate), sodium 2,3,-bis(2-methoxy-4-nitro-5-sulphophenyl)-5-[(phenylamino)-carbonyl]-2H-tetrazolium inner salt

N812-100MGEA 0 EUR
N812-100MG
XTT Natrium Salz, hochrein
XTT sodium salt
Formel: C₂₂H₁₇N₇O₁₃S₂Na
Molecular Weight: 673.52 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00083517
CAS-Nummer: 111072-31-2

Bestellen

Spezifikation Testergebnisse

Emax (285nm) 14800 - 16800
Appearance Yellow powder
Water (KF) ≤ 15 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR