VWR® EZ-Vision®, DNA Ladder, 100 bp
Lieferant: VWR Chemicals
N856-300ULEA
177
EUR
N856-300UL
VWR® EZ-Vision®, DNA Ladder, 100 bp
Elektrophorese-Reagenzien
Molekulargewichtsmarker
EZ-Vision® 100 bp DNA ladder is a pre-stained molecular weight ladder in loading buffer that allows immediate visualisation of DNA ladder post-electrophoresis.
- 13 fragments ranging from 100 to 3000 bp with reference bands for easy identification
- 'Load-and-go' formula with instant visualisation
- Eliminates the use of ethidium bromide