3-[(3-Cholamidopropyl)dimethylammonio]-1-propansulfonat (CHAPS) 10% in wässriger Lösung, Reagenzqualität
Lieferant: VWR Chemicals
|
Achtung
|
Synonyme:
CHAPS
N907-1LEA
663
EUR
N907-1L
3-[(3-Cholamidopropyl)dimethylammonio]-1-propansulfonat (CHAPS) 10% in wässriger Lösung, Reagenzqualität
3-[(3-Cholamidopropyl)dimethylammonio]-1-propansulfonat (CHAPS)
Formel:
C₃₂H₅₈N₂O₇S Molecular Weight: 614,89 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00081083 CAS-Nummer: 75621-03-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...