Total organic carbon (TOC) vial, Nalgene®
Lieferant: Thermo Fisher Scientific
1000-0000
NALG1000-0000EA 191 EUR
NALG1000-0000
Total organic carbon (TOC) vial, Nalgene®
Vials
TOC-certified vials are low-level–certified. Several sizes of containers are available, including the popular autosampler vials cleaned, certified and ready to use. Simplify and reduce the cost of cleaning validations.
  • Low background is perfect for preparation and storage of standards
  • Off-line and grab sampling of high-purity water