Beakers, low form, PPCO, set, Nalgene®
Lieferant: Thermo Fisher Scientific
1201-1234
NALG1201-1234EA 56 EUR
NALG1201-1234
Beakers, low form, PPCO, set, Nalgene®
Becher
A convenient and cost effective way to get started setting up a new lab, assembling student lab kits, or refreshing an existing lab.
  • No drip single-spout design provides precision pouring control
  • With ribs for easy stacking
  • Impact resistant at temperatures above –40 °C
  • Autoclavable/graduated compliance
  • Meets ISO/DIN 7056 International Standards for laboratory plasticware beakers and 21 CFR Part 177,1520 for food/beverage use
Silk screened with graduations, size code, resin code, catalog number, max/ temperature and 'No Flame' symbol for easy identification and safe use.
Standards: Meets ISO/DIN 7056 International Standards for laboratory plasticware beakers and 21 CFR Part 177.1520 for food/beverage use.
Bestellinformation: Includes seven beakers, one each size: 50, 100, 150, 250, 400, 600 and 1000 ml.