Bottles, narrow neck, round, LDPE, Boston type, with screw caps, Nalgene®
Lieferant: Thermo Fisher Scientific
312003-0001 312003-0002 312003-0004 312003-0008 312003-0016 312003-0032
NALG312003-0001EA 1030 EUR
NALG312003-0001 NALG312003-0002 NALG312003-0004 NALG312003-0008 NALG312003-0016 NALG312003-0032
Bottles, narrow neck, round, LDPE, Boston type, with screw caps, Nalgene®
Flaschen Kunststoffflaschen
Boston round narrow-mouth LDPE bottles with closure, constructed from natural low-density polyethylene.
  • Good chemical resistance, heat resistant up to 80 °C
  • Well suited for trace analysis
  • More flexible and impact resistant than HDPE bottles, leakproof
  • Non sterile
Ideal for storing and shipping liquids.
Uniform walls that resist splitting and puncturing. Leakproof when used with Nalgene closures.
Standards: Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.