Tamper evident closures for PET/PETG square media bottles, HDPE
Lieferant: Thermo Fisher Scientific
342141-0384
NALG342141-0384EA 206 EUR
NALG342141-0384
Tamper evident closures for PET/PETG square media bottles, HDPE
Verschlüsse Flaschenverschlüsse
Closure, tamper evident, specially designed for use with PET/PETG square media bottles.
  • Natural HDPE closures are ADCF (Animal Derived Component-Free)
  • Fits Nalgene PET and PETG Media Bottles with 38-430 neck finish and leakproof guarantee
  • Easy application and breaking of the tamper evident seal ring
  • Gamma irradiated to provide 10⁻⁶ Sterility Assurance Level (SAL)
  • Flat top surface for possible printing or labelling
Verpackung: Closures are sterile and bulk packed.