Aräometer-Zylinder, Nalgene®
Lieferant: Thermo Fisher Scientific
6230-0500
NALG6230-0500EA 38.9 EUR
NALG6230-0500
Aräometer-Zylinder, Nalgene®
Zylinder Aräometerzylinder
Made of PMP, clear.
  • Superior chemical resistance
  • With broad base
  • Good temperature resistance
PMP protect against breakage with the hydrometer cylinder. Lighter in weight than glass. This transparent, autoclavable cylinder can also be used as a lactometer cylinder.