Laborbücher, Nalgene®
Lieferant: Thermo Fisher Scientific
6501-1000
NALG6501-1000EA 68.4 EUR
NALG6501-1000
Laborbücher, Nalgene®
Notizbücher
Polyethylene cover. Specially designed for process record keeping and ideal for research laboratories in securing patent protection. Featuring 184 high quality acid-free, paper pages with ¹/₄" grid lines. The 8¹/₂×11" pages are permanently bound and sewn.
  • Page format includes place for preparer and witness signatures to aid in securing patent protection Burgundy cover is water resistant
Format: A4.
Colour: burgundy.