Schläuche, FEP, Nalgene®
Lieferant: Thermo Fisher Scientific
8050-0125 8050-0187 8050-0250 8050-0310 8050-0375 8050-0437 8050-0500 8050-0562
NALG8050-0125EA 105 EUR
NALG8050-0125 NALG8050-0187 NALG8050-0250 NALG8050-0310 NALG8050-0375 NALG8050-0437 NALG8050-0500 NALG8050-0562
Schläuche, FEP, Nalgene®
Schläuche Transferschlauch
FEP, translucent.
  • Semi-rigid flexibility
  • Use for demanding lab applications where binding/adsorption is a problem
  • Superior non-stick characteristics eases movement of viscous, aggressive compounds
  • Can be sterilized by autoclaving, gas, dry heat and disinfectants
Tubing is rated excellent for: acids, aliphatic alcohols, aldehydes, bases, esters, hydrocarbons, ketones and strong oxidizing agents.
Standards: Meets FDA 21 CFR 177.1550 for food contact.