Schläuche, PFA, Nalgene®
Lieferant: Thermo Fisher Scientific
8051-0125 8051-0187 8051-0250 8051-0375 8051-0500
NALG8051-0125EA 148 EUR
NALG8051-0125 NALG8051-0187 NALG8051-0250 NALG8051-0375 NALG8051-0500
Schläuche, PFA, Nalgene®
Schläuche Transferschlauch
Made of PFA, translucent.
  • Semi-rigid with very high chemical resistance
  • Wide temperature range: –270 to 260 °C
  • Durometer hardness: Shore D, 55
  • Very low water absorption
For low-pressure use with mechanical, compression-type fittings. Can be sterilized by autoclaving, gas, dry heat, and disinfectants. Do not sterilize by radiation.
Standards: Resin complies with USDA regulations.