Non-phthalate PVC vacuum tubing, Nalgene™
Lieferant: Thermo Fisher Scientific
8702-0055 8702-0065 8702-1065 8702-0145 8702-1145 8702-0210 8702-1210
NALG8702-0055EA 239 EUR
NALG8702-0055 NALG8702-0065 NALG8702-1065 NALG8702-0145 NALG8702-1145 NALG8702-0210 NALG8702-1210
Non-phthalate PVC vacuum tubing, Nalgene™
Schläuche Vakuum-Schläuche und Druck-Schläuche
Nalgene™ lab-grade, autoclavable clear tubing sets the standard for purity and quality, made from highest-grade resins, free from ortho-phthalate plasticisers.
  • Compare to Tygon® E-3603 tubing
  • Marked with Int.Ø and regulatory compliance as well as at 305 mm intervals for easy identification and measurement
  • Durometer hardness is shore A, 60
  • Thick-walled PVC tubing withstands a full vacuum of 760 mm Hg at room temperature
Nalgene™ Non-phthalate PVC vacuum tubing is manufactured from the highest-grade resin materials and performs like classic Nalgene™ 180More Product Information